Assay Details
| Gene Symbol: 4930481A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027938.2 | 3 - 4 | 551 | 87 | ||
| GenBank mRNA | BM937611.1 | - | 3 - 4 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930481A15Rik| Gene Symbol: 4930481A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027938.2 | 3 - 4 | 551 | 87 | ||
| GenBank mRNA | BM937611.1 | - | 3 - 4 | 87 | ||
Set Membership: |
|