Assay Details
| Gene Symbol: Slc3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161413.1 | NP_001154885.1 | 1 - 2 | 382 | 90 | |
| GenBank mRNA | AK161280.1 | - | 1 - 2 | 383 | 90 | |
| AK186746.1 | - | 1 - 2 | 53 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK161280.1 transcripts.
Set Membership: |
|

Slc3a2
Genomic Map