Assay Details
| Gene Symbol: Msn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010833.2 | NP_034963.2 | 1 - 2 | 203 | 63 | |
| GenBank mRNA | AK031171.1 | - | 1 - 2 | 193 | 63 | |
| AK087001.1 | - | 1 - 2 | 152 | 63 | ||
| AK088336.1 | - | 1 - 2 | 151 | 63 | ||
| AK133819.1 | - | 1 - 2 | 207 | 63 | ||
| AK145746.1 | - | 1 - 2 | 207 | 63 | ||
| AK150869.1 | - | 1 - 2 | 206 | 63 | ||
| AK153210.1 | - | 1 - 2 | 206 | 63 | ||
| AK157701.1 | - | 1 - 2 | 158 | 63 | ||
| BC003474.1 | - | 1 - 2 | 194 | 63 | ||
| BC047366.1 | - | 1 - 2 | 197 | 3711212 | 63 | |
| BY336882.1 | - | 1 - 2 | 203 | 63 | ||
| S47577.1 | - | 1 - 2 | 188 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK031171.1, AK088336.1, AK133819.1, AK145746.1, AK150869.1, AK153210.1, AK157701.1, BC003474.1, BC047366.1, NM_010833.2, S47577.1 transcripts.
Set Membership: |
|

Msn
Genomic Map