Assay Details
| Gene Symbol: Adhfe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175236.4 | NP_780445.1 | 4 - 5 | 231 | 208 | |
| NR_027664.1 | 3 - 4 | 190 | 167 | |||
| GenBank mRNA | AK018241.1 | - | 4 - 5 | 205 | 208 | |
| AK038853.1 | - | 4 - 5 | 198 | 208 | ||
| AK046780.1 | - | 4 - 5 | 157 | 208 | ||
| AK050178.1 | - | 3 - 4 | 155 | 167 | ||
| AK084104.1 | - | 158 | 208 | |||
| BC026584.1 | - | 4 - 5 | 189 | 4972323 | 208 | |
| CA467848.1 | - | 4 - 5 | 270 | 208 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK050178.1 transcripts.
Set Membership: |
|

Adhfe1
Genomic Map