Assay Details
| Gene Symbol: D130009I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015593.2 | 2 - 3 | 998 | 147 | ||
| GenBank mRNA | AK051495.1 | - | 2 - 3 | 998 | 147 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ring finger protein 219 | Rnf219 |
2610206B13Rik, 2810449K13Rik, AI451544 |
-207.4kb | Anti-sense | Chr. 14 - 104477534 - 104522711 on NCBI Build GRCm38 |
| Downstream | predicted gene 17066 | Gm17066 | 221.4kb | Anti-sense | Chr. 14 - 105106728 - 105114676 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

D130009I18Rik
Genomic Map