Assay Details
| Gene Symbol: Lhcgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013582.2 | NP_038610.1 | 3 - 4 | 329 | 84 | |
| GenBank mRNA | AK030325.1 | - | 3 - 4 | 344 | 84 | |
| AK054331.1 | - | 2 - 3 | 166 | 84 | ||
| M81310.1 | - | 3 - 4 | 329 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lhcgr
Genomic Map