Assay Details
| Gene Symbol: Gtf2a1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023630.2 | NP_076119.2 | 4 - 5 | 345 | 90 | |
| GenBank mRNA | AF250835.1 | - | 4 - 5 | 313 | 90 | |
| AK005881.1 | - | 4 - 5 | 320 | 90 | ||
| BC050756.1 | - | 4 - 5 | 328 | 6741860 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2a1l
Genomic Map