Assay Details
| Gene Symbol: Gtf2a1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023630.2 | NP_076119.2 | 7 - 8 | 1261 | 69 | |
| GenBank mRNA | AF250835.1 | - | 7 - 8 | 1229 | 69 | |
| AK005881.1 | - | 7 - 8 | 1236 | 69 | ||
| BC050756.1 | - | 7 - 8 | 1244 | 6741860 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2a1l
Genomic Map