Assay Details
| Gene Symbol: A230004M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK049168.1 | - | 2 - 3 | 342 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cyclin G1 | Ccng1 | AI314029 | -958.7kb | Anti-sense | Chr. 11 - 40748552 - 40755315 on NCBI Build GRCm38 |
| Downstream |
gamma-aminobutyric acid (GABA) A receptor, subunit gamma 2 |
Gabrg2 |
GABAA-R, Gabrg-2, gamma2 |
105.8kb | Anti-sense | Chr. 11 - 41910190 - 42000840 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

A230004M16Rik
Genomic Map