Assay Details
| Gene Symbol: Ahsa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290654.1 | NP_001277583.1 | 6 - 7 | 856 | 117 | |
| NM_001290655.1 | NP_001277584.1 | 6 - 7 | 861 | 117 | ||
| NM_172391.4 | NP_765979.3 | 6 - 7 | 861 | 117 | ||
| GenBank mRNA | AK033696.1 | - | 6 - 7 | 769 | 117 | |
| AK035941.1 | - | 6 - 7 | 634 | 117 | ||
| AK093945.1 | - | 6 - 7 | 768 | 117 | ||
| AK137116.1 | - | 6 - 7 | 772 | 117 | ||
| AK154196.1 | - | 6 - 7 | 772 | 117 | ||
| AK155265.1 | - | 6 - 7 | 772 | 117 | ||
| BC038397.1 | - | 6 - 7 | 747 | 3676293 | 117 | |
| BC052829.1 | - | 6 - 7 | 779 | 30040034 | 117 | |
| BC055873.1 | - | 6 - 7 | 723 | 117 | ||
| BC064012.1 | - | 6 - 7 | 681 | 30135289 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ahsa2
Genomic Map