Assay Details
| Gene Symbol: Abcb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009592.1 | NP_033722.1 | 14 - 15 | 1962 | 133 | |
| GenBank mRNA | AK040069.1 | - | 14 - 15 | 1959 | 133 | |
| AK084853.1 | - | 14 - 15 | 1953 | 132 | ||
| AK151967.1 | - | 14 - 15 | 1948 | 133 | ||
| AK152535.1 | - | 14 - 15 | 1948 | 133 | ||
| AK152816.1 | - | 14 - 15 | 1948 | 133 | ||
| AK152946.1 | - | 14 - 15 | 1950 | 133 | ||
| BC035534.1 | - | 11 - 12 | 1591 | 133 | ||
| BC151037.1 | - | 14 - 15 | 1962 | 9087950 | 133 | |
| U43892.1 | - | 13 - 14 | 1766 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb7
Genomic Map