Assay Details
| Gene Symbol: Abcb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009592.1 | NP_033722.1 | 15 - 16 | 2067 | 93 | |
| GenBank mRNA | AK040069.1 | - | 15 - 16 | 2064 | 93 | |
| AK151967.1 | - | 15 - 16 | 2053 | 93 | ||
| AK152535.1 | - | 15 - 16 | 2053 | 93 | ||
| AK152816.1 | - | 15 - 16 | 2053 | 93 | ||
| AK152946.1 | - | 15 - 16 | 2055 | 93 | ||
| BC035534.1 | - | 12 - 13 | 1696 | 93 | ||
| BC151037.1 | - | 15 - 16 | 2067 | 9087950 | 93 | |
| U43892.1 | - | 14 - 15 | 1871 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb7
Genomic Map