Assay Details
| Gene Symbol: Abcb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009592.1 | NP_033722.1 | 3 - 4 | 349 | 142 | |
| GenBank mRNA | AK040069.1 | - | 3 - 4 | 347 | 142 | |
| AK084853.1 | - | 3 - 4 | 340 | 142 | ||
| AK151967.1 | - | 3 - 4 | 335 | 142 | ||
| AK152535.1 | - | 3 - 4 | 335 | 142 | ||
| AK152816.1 | - | 3 - 4 | 335 | 142 | ||
| AK152946.1 | - | 3 - 4 | 336 | 142 | ||
| BB612154.1 | - | 3 - 4 | 349 | 142 | ||
| BC151037.1 | - | 3 - 4 | 349 | 9087950 | 142 | |
| U43892.1 | - | 2 - 3 | 153 | 142 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb7
Genomic Map