Assay Details
| Gene Symbol: Abcb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009592.1 | NP_033722.1 | 4 - 5 | 481 | 95 | |
| GenBank mRNA | AK040069.1 | - | 4 - 5 | 479 | 95 | |
| AK084853.1 | - | 4 - 5 | 472 | 95 | ||
| AK151967.1 | - | 4 - 5 | 467 | 95 | ||
| AK152535.1 | - | 4 - 5 | 467 | 95 | ||
| AK152816.1 | - | 4 - 5 | 467 | 95 | ||
| AK152946.1 | - | 4 - 5 | 468 | 95 | ||
| BB612154.1 | - | 4 - 5 | 95 | |||
| BC035534.1 | - | 1 - 2 | 110 | 95 | ||
| BC151037.1 | - | 4 - 5 | 481 | 9087950 | 95 | |
| U43892.1 | - | 3 - 4 | 285 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb7
Genomic Map