Assay Details
| Gene Symbol: Abcb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009592.1 | NP_033722.1 | 5 - 6 | 600 | 71 | |
| GenBank mRNA | AK040069.1 | - | 5 - 6 | 598 | 71 | |
| AK084853.1 | - | 5 - 6 | 591 | 71 | ||
| AK151967.1 | - | 5 - 6 | 586 | 71 | ||
| AK152535.1 | - | 5 - 6 | 586 | 71 | ||
| AK152816.1 | - | 5 - 6 | 586 | 71 | ||
| AK152946.1 | - | 5 - 6 | 587 | 71 | ||
| BC035534.1 | - | 2 - 3 | 229 | 71 | ||
| BC151037.1 | - | 5 - 6 | 600 | 9087950 | 71 | |
| U43892.1 | - | 4 - 5 | 404 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb7
Genomic Map