Assay Details
| Gene Symbol: Abcb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009592.1 | NP_033722.1 | 6 - 7 | 876 | 76 | |
| GenBank mRNA | AK040069.1 | - | 6 - 7 | 873 | 76 | |
| AK084853.1 | - | 6 - 7 | 867 | 76 | ||
| AK151967.1 | - | 6 - 7 | 862 | 76 | ||
| AK152535.1 | - | 6 - 7 | 862 | 76 | ||
| AK152816.1 | - | 6 - 7 | 862 | 76 | ||
| AK152946.1 | - | 6 - 7 | 863 | 76 | ||
| BC035534.1 | - | 3 - 4 | 505 | 76 | ||
| BC151037.1 | - | 6 - 7 | 876 | 9087950 | 76 | |
| U43892.1 | - | 5 - 6 | 680 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb7
Genomic Map