Assay Details
| Gene Symbol: Gm28154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034407.1 | - | 1 - 2 | 3674 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | peptidase inhibitor 15 | Pi15 |
P24TI, P25TI, PI-15, SugarCrisp |
-6.1kb | Anti-sense | Chr. 1 - 17601901 - 17630939 on NCBI Build GRCm38 |
| Downstream | predicted gene 16070 | Gm16070 | OTTMUSG00000029756 | 34.9kb | Sense | Chr. 1 - 17676027 - 17727646 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm28154
Genomic Map