Assay Details
| Gene Symbol: 2610203C22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015470.1 | 4 - 5 | 294 | 161 | ||
| GenBank mRNA | BC037765.1 | - | 4 - 5 | 294 | 161 | |
| BC064105.1 | - | 4 - 5 | 294 | 161 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RRS1 ribosome biogenesis regulator homolog (S. cerevisiae) | Rrs1 |
5730466A07Rik, AA415037, D1Ertd701e, Rrr |
-13.4kb | Anti-sense | Chr. 1 - 9545408 - 9547455 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 3110035E14 gene | 3110035E14Rik |
AI415019, AV164974, R75066 |
37.4kb | Anti-sense | Chr. 1 - 9601267 - 9627142 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

2610203C22Rik
Genomic Map