Assay Details
| Gene Symbol: Abcg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286005.1 | NP_001272934.1 | 4 - 5 | 551 | 73 | |
| NM_026180.3 | NP_080456.1 | 5 - 6 | 803 | 73 | ||
| NR_104382.1 | 5 - 6 | 800 | 73 | |||
| GenBank mRNA | AF324495.1 | - | 5 - 6 | 793 | 73 | |
| AK004871.1 | - | 5 - 6 | 760 | 73 | ||
| AK050938.1 | - | 4 - 5 | 553 | 73 | ||
| AY196215.1 | - | 5 - 6 | 790 | 73 | ||
| AY196216.1 | - | 5 - 6 | 790 | 73 | ||
| BC138484.1 | - | 5 - 6 | 767 | 8861504 | 73 | |
| BC145293.1 | - | 5 - 6 | 767 | 9053847 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcg8
Genomic Map