Assay Details
| Gene Symbol: Prepl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163622.1 | NP_001157094.1 | 7 - 8 | 1443 | 139 | |
| NM_001163623.1 | NP_001157095.1 | 6 - 7 | 907 | 139 | ||
| NM_001163624.1 | NP_001157096.1 | 6 - 7 | 775 | 139 | ||
| NM_145984.3 | NP_666096.3 | 7 - 8 | 1007 | 139 | ||
| GenBank mRNA | AK028877.1 | - | 7 - 8 | 1401 | 139 | |
| AK050878.1 | - | 6 - 7 | 769 | 139 | ||
| AK083070.1 | - | 7 - 8 | 948 | 139 | ||
| AK129142.1 | - | 1 - 2 | 861 | 139 | ||
| BC004612.1 | - | 6 - 7 | 861 | 3585402 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prepl
Genomic Map