Assay Details
| Gene Symbol: Prepl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163622.1 | NP_001157094.1 | 9 - 10 | 1832 | 86 | |
| NM_001163623.1 | NP_001157095.1 | 8 - 9 | 1296 | 86 | ||
| NM_001163624.1 | NP_001157096.1 | 8 - 9 | 1164 | 86 | ||
| NM_145984.3 | NP_666096.3 | 9 - 10 | 1396 | 86 | ||
| GenBank mRNA | AK028877.1 | - | 9 - 10 | 1790 | 86 | |
| AK050878.1 | - | 8 - 9 | 1158 | 86 | ||
| AK083070.1 | - | 9 - 10 | 1337 | 86 | ||
| AK129142.1 | - | 3 - 4 | 1250 | 86 | ||
| BC004612.1 | - | 8 - 9 | 1250 | 3585402 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prepl
Genomic Map