Assay Details
| Gene Symbol: Sntg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290390.1 | NP_001277319.1 | 16 - 17 | 2936 | 142 | |
| NM_001290392.1 | NP_001277321.1 | 13 - 14 | 2671 | 142 | ||
| NM_001290393.1 | NP_001277322.1 | 15 - 16 | 2885 | 142 | ||
| NM_027671.5 | NP_081947.2 | 16 - 17 | 2939 | 142 | ||
| GenBank mRNA | AF367759.1 | - | 15 - 16 | 1591 | 142 | |
| AK016927.1 | - | 2 - 3 | 228 | 142 | ||
| AK038698.1 | - | 13 - 14 | 2672 | 142 | ||
| AK080034.1 | - | 16 - 17 | 2301 | 142 | ||
| AK083241.1 | - | 16 - 17 | 1924 | 142 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sntg1
Genomic Map