Assay Details
| Gene Symbol: Sntg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290390.1 | NP_001277319.1 | 7 - 8 | 2192 | 89 | |
| NM_001290392.1 | NP_001277321.1 | 6 - 7 | 2138 | 89 | ||
| NM_001290393.1 | NP_001277322.1 | 6 - 7 | 2138 | 89 | ||
| NM_027671.5 | NP_081947.2 | 7 - 8 | 2192 | 89 | ||
| GenBank mRNA | AF367759.1 | - | 6 - 7 | 844 | 89 | |
| AK038698.1 | - | 6 - 7 | 2139 | 89 | ||
| AK045676.1 | - | 6 - 7 | 1523 | 89 | ||
| AK080034.1 | - | 7 - 8 | 1554 | 89 | ||
| AK083241.1 | - | 7 - 8 | 1180 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK038698.1, NM_001290392.1 transcripts.
Set Membership: |
|

Sntg1
Genomic Map