Assay Details
| Gene Symbol: Sntg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290390.1 | NP_001277319.1 | 10 - 11 | 2334 | 107 | |
| NM_001290392.1 | NP_001277321.1 | 9 - 10 | 2283 | 107 | ||
| NM_001290393.1 | NP_001277322.1 | 9 - 10 | 2283 | 107 | ||
| NM_027671.5 | NP_081947.2 | 10 - 11 | 2337 | 107 | ||
| GenBank mRNA | AF367759.1 | - | 9 - 10 | 989 | 107 | |
| AK038698.1 | - | 9 - 10 | 2284 | 107 | ||
| AK080034.1 | - | 10 - 11 | 1699 | 107 | ||
| AK083241.1 | - | 10 - 11 | 1322 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK038698.1, NM_001290392.1 transcripts.
Set Membership: |
|

Sntg1
Genomic Map