Assay Details
| Gene Symbol: Nckipsd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030729.4 | NP_109654.2 | 11 - 12 | 1873 | 128 | |
| GenBank mRNA | AF130313.2 | - | 11 - 12 | 1769 | 128 | |
| AK134725.1 | - | 11 - 12 | 1855 | 128 | ||
| BC064818.1 | - | 11 - 12 | 1873 | 30292108 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nckipsd
Genomic Map