Assay Details
| Gene Symbol: Nckipsd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030729.4 | NP_109654.2 | 6 - 7 | 1334 | 69 | |
| GenBank mRNA | AF130313.2 | - | 6 - 7 | 1230 | 69 | |
| AK134725.1 | - | 6 - 7 | 1316 | 69 | ||
| BC064818.1 | - | 6 - 7 | 1334 | 30292108 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nckipsd
Genomic Map