Assay Details
| Gene Symbol: P4htm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028944.3 | NP_083220.3 | 2 - 3 | 476 | 79 | |
| GenBank mRNA | AK016685.1 | - | 2 - 3 | 450 | 79 | |
| AK038927.1 | - | 2 - 3 | 458 | 79 | ||
| AK046783.1 | - | 2 - 3 | 488 | 79 | ||
| AK047714.1 | - | 2 - 3 | 476 | 79 | ||
| AK161992.1 | - | 2 - 3 | 452 | 79 | ||
| BC111871.1 | - | 2 - 3 | 476 | 40057593 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

P4htm
Genomic Map