Assay Details
| Gene Symbol: Pfkfb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173019.5 | NP_766607.3 | 4 - 5 | 490 | 66 | |
| GenBank mRNA | AK155172.1 | - | 4 - 5 | 428 | 66 | |
| AY646092.2 | - | 4 - 5 | 376 | 66 | ||
| AY707862.1 | - | 4 - 5 | 393 | 66 | ||
| AY756065.1 | - | 4 - 5 | 376 | 66 | ||
| AY756066.1 | - | 4 - 5 | 376 | 66 | ||
| AY756067.1 | - | 4 - 5 | 376 | 66 | ||
| BC057594.1 | - | 4 - 5 | 442 | 6810667 | 66 | |
| CA316033.1 | - | 4 - 5 | 442 | 66 | ||
| DQ350638.1 | - | 5 - 6 | 462 | 66 | ||
| EU571215.1 | - | 4 - 5 | 490 | 66 | ||
| EU571216.1 | - | 6 - 7 | 649 | 66 | ||
| EU571217.1 | - | 4 - 5 | 617 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY756066.1, EU571217.1 transcripts.
Set Membership: |
|

Pfkfb4
Genomic Map