Assay Details
| Gene Symbol: Pfkfb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173019.5 | NP_766607.3 | 9 - 10 | 1101 | 73 | |
| GenBank mRNA | AK155172.1 | - | 9 - 10 | 1039 | 73 | |
| AY646092.2 | - | 9 - 10 | 987 | 73 | ||
| AY707862.1 | - | 9 - 10 | 1004 | 73 | ||
| AY756065.1 | - | 9 - 10 | 1094 | 73 | ||
| AY756066.1 | - | 9 - 10 | 1094 | 73 | ||
| AY756067.1 | - | 9 - 10 | 987 | 73 | ||
| BC057594.1 | - | 9 - 10 | 1053 | 6810667 | 73 | |
| DQ350638.1 | - | 10 - 11 | 1073 | 73 | ||
| EU571215.1 | - | 9 - 10 | 1101 | 73 | ||
| EU571216.1 | - | 11 - 12 | 1260 | 73 | ||
| EU571217.1 | - | 9 - 10 | 1228 | 73 | ||
More Information
Set Membership: |
|

Pfkfb4
Genomic Map