Assay Details
| Gene Symbol: Prkar2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008924.2 | NP_032950.1 | 5 - 6 | 845 | 129 | |
| GenBank mRNA | AF533977.1 | - | 5 - 6 | 845 | 129 | |
| AK132728.1 | - | 5 - 6 | 784 | 129 | ||
| AK132824.1 | - | 5 - 6 | 776 | 129 | ||
| BC075623.1 | - | 5 - 6 | 816 | 6854057 | 129 | |
| BC080276.1 | - | 5 - 6 | 839 | 30354403 | 129 | |
| J02935.1 | - | 5 - 6 | 528 | 129 | ||
More Information
Set Membership: |
|

Prkar2a
Genomic Map