Assay Details
| Gene Symbol: Dalrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026378.3 | NP_080654.3 | 10 - 11 | 1445 | 113 | |
| GenBank mRNA | AK002439.1 | - | 10 - 11 | 1441 | 113 | |
| AK003374.1 | - | 4 - 5 | 392 | 113 | ||
| AK149963.1 | - | 10 - 11 | 1446 | 113 | ||
| AK169405.1 | - | 10 - 11 | 1443 | 113 | ||
| AK171818.1 | - | 10 - 11 | 1446 | 113 | ||
| AK185660.1 | - | 1 - 2 | 38 | 113 | ||
| BC013483.1 | - | 10 - 11 | 1426 | 4242135 | 113 | |
| BC058774.1 | - | 8 - 9 | 844 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26815514 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26815513 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26815515 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26815516 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26815512 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158061 | BC013483.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158061 | AK169405.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158061 | AK171818.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158061 | AK149963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158061 | AK195213.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158061 | BC013483.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158061 | AK169405.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158061 | AK171818.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158061 | AK149963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158061 | AK195213.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 679405 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855125 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125508 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158061 | BC013483.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158061 | AK169405.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158061 | AK171818.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158061 | AK149963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158061 | AK195213.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158061 | BC013483.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158061 | AK169405.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158061 | AK171818.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158061 | AK149963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158061 | AK195213.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815519 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125508 | - | - |
| Whole Mouse Genome G4122A | A_52_P338199 | BC013483.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P338199 | BC058774.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P338199 | AK169405.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P338199 | AK171818.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P338199 | AK149963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158061 | BC013483.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158061 | AK169405.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158061 | AK171818.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158061 | AK149963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158061 | AK195213.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158061 | BC013483.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158061 | AK169405.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158061 | AK171818.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158061 | AK149963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158061 | AK195213.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101015 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125508 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07648896 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26815512 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26815515 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26815516 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26815514 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26815513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07648866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07648896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26815527 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10589073 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589063 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589065 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589070 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589071 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589066 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589072 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589069 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423894_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10589061 | - | - |
| Mouse Genome 430 2.0 Array | 1423894_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423894_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1447879_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1447879_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67789_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67789_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900011473.mm.1 | NM_026378.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011473.mm.1 | NM_026378.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011473.mm.1 | TC0900001416.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900011474.mm.1 | NM_026378.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011474.mm.1 | NM_026378.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011474.mm.1 | TC0900001416.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423894_PM_a_at | - | - |
| Mouse Expression Set 430 | 1423894_a_at | - | - |
| Mouse Expression Set 430 | 1447879_PM_x_at | - | - |
| Mouse Expression Set 430 | 1447879_x_at | - | - |
| Murine Genome U74v2 Set | 161766_i_at | - | - |
| Murine Genome U74v2 Set | 161766_PM_i_at | - | - |
| Murine Genome U74v2 Set | 160346_at | - | - |
| Murine Genome U74v2 Set | 160346_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423894_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423894_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423894_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423894_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6900195 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006900195 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2644238 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2644238 | - | - |
| mouse-6 v1.1 (Illumina) | scl36462.6_134-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6900195 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36462.6_134-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900195 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2644238 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2644238 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36462.6_134-S | - | - |

Dalrd3
Genomic Map