Assay Details
| Gene Symbol: Dalrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026378.3 | NP_080654.3 | 2 - 3 | 481 | 112 | |
| GenBank mRNA | AK002439.1 | - | 2 - 3 | 478 | 112 | |
| AK149963.1 | - | 2 - 3 | 482 | 112 | ||
| AK169405.1 | - | 2 - 3 | 479 | 112 | ||
| AK171818.1 | - | 2 - 3 | 482 | 112 | ||
| BC013483.1 | - | 2 - 3 | 462 | 4242135 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002439.1 transcripts.
Set Membership: |
|

Dalrd3
Genomic Map