Assay Details
| Gene Symbol: Dalrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026378.3 | NP_080654.3 | 4 - 5 | 811 | 114 | |
| GenBank mRNA | AK149963.1 | - | 4 - 5 | 812 | 114 | |
| AK169405.1 | - | 4 - 5 | 809 | 114 | ||
| AK171818.1 | - | 4 - 5 | 812 | 114 | ||
| BC013483.1 | - | 4 - 5 | 792 | 4242135 | 114 | |
| BC058774.1 | - | 2 - 3 | 210 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dalrd3
Genomic Map