Assay Details
| Gene Symbol: Dalrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026378.3 | NP_080654.3 | 6 - 7 | 1018 | 86 | |
| GenBank mRNA | AK002439.1 | - | 6 - 7 | 1014 | 86 | |
| AK149963.1 | - | 6 - 7 | 1019 | 86 | ||
| AK169405.1 | - | 6 - 7 | 1016 | 86 | ||
| AK171818.1 | - | 6 - 7 | 1019 | 86 | ||
| BC013483.1 | - | 6 - 7 | 999 | 4242135 | 86 | |
| BC058774.1 | - | 4 - 5 | 417 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002439.1 transcripts.
Set Membership: |
|

Dalrd3
Genomic Map