Assay Details
| Gene Symbol: Msn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010833.2 | NP_034963.2 | 4 - 5 | 655 | 66 | |
| GenBank mRNA | AK088336.1 | - | 4 - 5 | 603 | 66 | |
| AK133819.1 | - | 4 - 5 | 659 | 66 | ||
| AK145746.1 | - | 4 - 5 | 659 | 66 | ||
| AK150869.1 | - | 4 - 5 | 658 | 66 | ||
| AK153210.1 | - | 4 - 5 | 658 | 66 | ||
| AK157701.1 | - | 4 - 5 | 610 | 66 | ||
| BC003474.1 | - | 4 - 5 | 646 | 66 | ||
| BC047366.1 | - | 4 - 5 | 649 | 3711212 | 66 | |
| M86390.1 | - | 3 - 4 | 435 | 66 | ||
| S47577.1 | - | 4 - 5 | 640 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msn
Genomic Map