Assay Details
| Gene Symbol: Fez2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285940.1 | NP_001272869.1 | 3 - 4 | 543 | 83 | |
| NM_001285946.1 | NP_001272875.1 | 3 - 4 | 543 | 83 | ||
| NM_001285949.1 | NP_001272878.1 | 3 - 4 | 543 | 83 | ||
| NM_199448.3 | NP_955519.1 | 3 - 4 | 543 | 83 | ||
| GenBank mRNA | AK033547.1 | - | 3 - 4 | 485 | 83 | |
| AK150241.1 | - | 3 - 4 | 509 | 83 | ||
| AK159545.1 | - | 3 - 4 | 496 | 83 | ||
| BC052366.1 | - | 3 - 4 | 522 | 83 | ||
| BC064130.1 | - | 3 - 4 | 449 | 83 | ||
| BC096619.1 | - | 3 - 4 | 488 | 6391451 | 83 | |
More Information
Set Membership: |
|

Fez2
Genomic Map