Assay Details
| Gene Symbol: Gpatch11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181649.6 | NP_857632.4 | 6 - 7 | 751 | 107 | |
| GenBank mRNA | AK148331.1 | - | 6 - 7 | 753 | 107 | |
| BC047998.1 | - | 6 - 7 | 665 | 6511097 | 107 | |
| BC092522.1 | - | 6 - 7 | 678 | 30648550 | 107 | |
| BC138310.1 | - | 6 - 7 | 646 | 8861330 | 107 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpatch11
Genomic Map