Assay Details
| Gene Symbol: Il20rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033543.3 | NP_001028715.2 | 5 - 6 | 813 | 54 | |
| GenBank mRNA | AK137306.1 | - | 1 - 2 | 665 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il20rb
Genomic Map