Assay Details
| Gene Symbol: A4gnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077424.2 | NP_001070892.1 | 1 - 2 | 103 | 66 | |
| GenBank mRNA | AK136425.1 | - | 1 - 2 | 104 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001077424.2 transcripts.
Set Membership: |
|

A4gnt
Genomic Map