Assay Details
| Gene Symbol: A4gnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077424.2 | NP_001070892.1 | 2 - 3 | 596 | 60 | |
| GenBank mRNA | AK136425.1 | - | 2 - 3 | 598 | 60 | |
| BC115614.1 | - | 1 - 2 | 487 | 40102739 | 60 | |
| BC115615.1 | - | 1 - 2 | 487 | 40102743 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P175634 | AK136425.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P175634 | BC115614.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P175634 | BC115615.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P175634 | NM_001077424.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P175634 | AK136425.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P175634 | BC115614.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P175634 | BC115615.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P175634 | NM_001077424.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 769481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341119 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349026 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818587 | AK076842.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818587 | XM_001473407.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818587 | XM_001475243.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57240 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P175634 | AK136425.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P175634 | BC115614.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P175634 | BC115615.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P175634 | NM_001077424.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P175634 | AK136425.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P175634 | BC115614.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P175634 | BC115615.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P175634 | NM_001077424.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P175634 | AK136425.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P175634 | BC115614.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P175634 | BC115615.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P175634 | NM_001077424.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P175634 | AK136425.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P175634 | BC115614.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P175634 | BC115615.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P175634 | NM_001077424.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P175634 | AK136425.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P175634 | BC115614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P175634 | BC115615.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P175634 | NM_001077424.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P175634 | AK136425.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P175634 | BC115614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P175634 | BC115615.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P175634 | NM_001077424.2 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26765121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07964744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07683709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07744040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08144243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08183976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07545977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07802306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07911263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07896680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26765153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07513760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07549764 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10588136 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588135 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588133 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588134 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458930_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10588132 | - | - |
| Mouse Genome 430 2.0 Array | 1458930_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458930_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900009793.mm.1 | NM_001077424.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900009793.mm.1 | NM_001077424.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900009793.mm.1 | TC0900001253.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900009794.mm.1 | NM_001077424.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900009794.mm.1 | NM_001077424.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900009794.mm.1 | TC0900001253.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1458930_at | - | - |
| Mouse Expression Set 430 | 1458930_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458930_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458930_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 882208 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219796 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2845412 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2845412 | - | - |

A4gnt
Genomic Map