Assay Details
| Gene Symbol: Dbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031403.3 | NP_113580.2 | 3 - 4 | 540 | 161 | |
| GenBank mRNA | AK050090.1 | - | 3 - 4 | 541 | 161 | |
| AK090351.1 | - | 3 - 4 | 516 | 161 | ||
| AK139631.1 | - | 3 - 4 | 522 | 161 | ||
| BC006661.1 | - | 3 - 4 | 535 | 3501390 | 161 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dbr1
Genomic Map