Assay Details
| Gene Symbol: Poc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK154163.1 | - | 1 - 2 | 150 | 65 | |
| BC034901.1 | - | 1 - 2 | 60 | 5255069 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK154163.1, BC034901.1 transcripts.
Set Membership: |
|

Poc1a
Genomic Map