Assay Details
| Gene Symbol: Poc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027354.2 | NP_081630.2 | 2 - 3 | 482 | 70 | |
| GenBank mRNA | AK154163.1 | - | 2 - 3 | 232 | 70 | |
| BC034901.1 | - | 2 - 3 | 142 | 5255069 | 70 | |
| CA557124.1 | - | 2 - 3 | 132 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Poc1a
Genomic Map