Assay Details
| Gene Symbol: Poc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027354.2 | NP_081630.2 | 6 - 7 | 1054 | 65 | |
| GenBank mRNA | AK011064.1 | - | 6 - 7 | 732 | 65 | |
| AK154163.1 | - | 6 - 7 | 804 | 65 | ||
| BC034901.1 | - | 6 - 7 | 714 | 5255069 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poc1a
Genomic Map