Assay Details
| Gene Symbol: Poc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027354.2 | NP_081630.2 | 8 - 9 | 1262 | 113 | |
| GenBank mRNA | AK011064.1 | - | 8 - 9 | 940 | 113 | |
| AK081217.1 | - | 447 | 113 | |||
| AK154163.1 | - | 8 - 9 | 1012 | 113 | ||
| BC034901.1 | - | 8 - 9 | 922 | 5255069 | 113 | |
| BE981233.1 | - | 3 - 4 | 113 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Poc1a
Genomic Map