Assay Details
| Gene Symbol: Poc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027354.2 | NP_081630.2 | 9 - 10 | 1354 | 89 | |
| GenBank mRNA | AK081217.1 | - | 539 | 89 | ||
| AK154163.1 | - | 9 - 10 | 1104 | 89 | ||
| BC034901.1 | - | 9 - 10 | 1014 | 5255069 | 89 | |
| BE981233.1 | - | 2 - 3 | 89 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Poc1a
Genomic Map