Assay Details
| Gene Symbol: Acy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276442.1 | NP_001263371.1 | 5 - 6 | 468 | 61 | |
| NM_025371.3 | NP_079647.1 | 5 - 6 | 472 | 61 | ||
| GenBank mRNA | AK002423.1 | - | 5 - 6 | 458 | 61 | |
| AK003703.1 | - | 5 - 6 | 473 | 61 | ||
| BC005631.1 | - | 5 - 6 | 476 | 3964138 | 61 | |
| CV560337.1 | - | 5 - 6 | 393 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acy1
Genomic Map