Assay Details
| Gene Symbol: Alas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291835.1 | NP_001278764.1 | 3 - 4 | 1012 | 86 | |
| NM_020559.2 | NP_065584.2 | 3 - 4 | 625 | 86 | ||
| GenBank mRNA | AK077152.1 | - | 2 - 3 | 316 | 86 | |
| AK133283.1 | - | 3 - 4 | 1013 | 86 | ||
| BC022110.1 | - | 3 - 4 | 527 | 5133249 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alas1
Genomic Map