Assay Details
| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011104.3 | NP_035234.1 | 2 - 3 | 1222 | 89 | |
| GenBank mRNA | AF028009.1 | - | 2 - 3 | 423 | 89 | |
| AF325507.1 | - | 2 - 3 | 414 | 89 | ||
| AK082438.1 | - | 2 - 3 | 1223 | 89 | ||
| AK085351.1 | - | 3 - 4 | 634 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkce
Genomic Map