Assay Details
| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011104.3 | NP_035234.1 | 9 - 10 | 2071 | 94 | |
| GenBank mRNA | AF028009.1 | - | 9 - 10 | 1272 | 94 | |
| AF325507.1 | - | 9 - 10 | 1263 | 94 | ||
| AK082438.1 | - | 9 - 10 | 2072 | 94 | ||
| AK085351.1 | - | 10 - 11 | 1483 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkce
Genomic Map